E.Z.N.A.® Universal Pathogen Kit, Omega Bio-Tek
Supplier: Omega Bio-Tek
E.Z.N.A®
D4035-01 D4035-01 D4035-00 D4035-00
CA75792-040EA 46.75 CAD
CA75792-040 75792-042 75792-040 CA75998-022
E.Z.N.A.® Universal Pathogen Kit, Omega Bio-Tek
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Universal Pathogen Kit for the rapid and reliable isolation of high quality host genomic DNA, as well as gram positive and negative bacterial DNA.
  • Elution volumes as low as 15 uL
  • Maintain higher nucleic acid concentrations
  • Rapid and reliable isolation of DNA
Universal Pathogen Kit, for the rapid and reliable isolation of high quality host genomic DNA, as well as gram positive and negative bacterial DNA. Elution volumes as low as 15 uL can be used to maintain higher nucleic acid concentrations.