Wire Gauze with Ceramic Center
Supplier: United Scientific Supplies
WGCC55-PK/12
WGCC66-PK/12
WGCC44-PK/12
WGCC66
WGCC55
470019-506PK
78.6
CAD
470019-506
470019-508
470019-504
470144-810
470144-812
Wire Gauze with Ceramic Center
Chemicals for Science Education
This durable gauze has a ceramic fiber center for flame protection and a longer life.
- Ceramic Fiber Center
- AWG No. 24 Galvanized Iron Wire
- Gauze Woven with Turned Edges to Prevent Fraying
- Organic Binder Included
The ceramic center is permanently enmeshed under high pressure to eliminate peeling.
The paper contains a small quantity of organic binder that temporarily produces a small flame and/or small amount of smoke when the gauze is first heated. The gauze woven with turned edges to prevent fraying.