Centurion™ 452 Series Impact Safety Goggles, 3M
Supplier: 3M
Centurion™
40301-00000-10 40301-00000-10
CA33001-599CS 127.41 CAD
33001-599 CA33001-599
Centurion™ 452 Series Impact Safety Goggles, 3M
Goggles
Centurion™ Safety Impact Goggle is a reliable choice in protective eyewear. Stable, sturdy construction will help protect wearer's eyes.
  • Wide-angle wraparound lens and frame for good peripheral vision
  • Direct venting system to help minimize fogging
  • Accommodates most prescription eyewear
Includes wide nasal flare for added comfort and a direct venting system to minimize fogging. All goggles feature a strap attachment system.
Certifications: Meets ANSI Z87.1-2010, CSA Z94.3-2007.