Water, Cell Culture Grade

Supplier: GROWCELLS

CCPW-2000 CCPW-010L CCPW-1000
10218-876CS 793.56 CAD
10218-876 10218-880 10218-882 10220-414 75792-294 75792-296
Water, Cell Culture Grade
Water

Distilled cell culture grade water.


  • Prepared by distillation
  • Steam sterilization in final packaging
  • 0.1 micron sterile filtration


Distilled cell culture grade water, <0.005EU/mL. Prepared by distillation, 0.1 micron sterile filtration and steam sterilization in final packaging.

Formula: H₂O
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR