Water, distilled

Supplier: GROWCELLS

Synonyms: Hydrogen oxide

SDW-2000 SDW-010L ISDW-010Q
10218-874CS 820.72 CAD
10218-874 10218-900 10218-902 10220-376
Water, distilled
Water

Used in the preparation of cell culture media and laboratory reagents.


  • Cell culture quality
  • Molecular biological grade
  • Prep laboratory reagents


Used in the preparation of cell culture media and laboratory reagents. Cell culture quality and molecular biological grade.


Ordering information: Water and buffers in IV bags are not injectable. They are not a medical device and are not to be used for intravenous use. They are for research only.

Formula: H₂O
MW: 18.02 g/mol
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR