VWR® Sampling Scoops, PS
Supplier: VWR International
10820-349 10820-351
10820-349EA 480.95 CAD
10820-349 10820-351
VWR® Sampling Scoops, PS
Scoops
Scoops are ideal for the sampling of powder, granules and pastes.
  • Individually packed, gamma radiation sterilized
Special handle construction and the slightly angled edge stands horizontal and can be laid down without the material trickling out.
Certifications: Production, assembling and packaging according to cleanroom Class 7 (10,000). Complies with food and FDA guidelines.