SteriPlast® Sampling Kit, Bürkle
Supplier: BURKLE INC
5378-8009 5378-8003
75836-568PK 113.62 CAD
75836-568 75836-624
SteriPlast® Sampling Kit, Bürkle
Scoops
The sampling set SteriPlast Kit is the ideal combination for sampling where purity and sterility is essential.
  • All parts are sterilized by gamma rays
  • Suitable for use with foodstuffs
  • Scoops or spatulas are cleanroom manufactured and individually packed
  • Sample bag SteriBag Premium has an integrated sealing and is liquid-tight
Unique tracking code on both the bag and the tear-off field guarantees complete documentation and traceability.
Delivery information: SteriPlast kit includes scoop 50 mL or spatula (150 mm insertion depth), and SteriBag Premium 300 mL.