HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid free acid ≥99% (by titrimetric analysis on dry substance), ULTROL®, Calbiochem®, Millipore®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ100

391338-100GM 391338-1KG 391338-25GM 391338-500GM 391338-5KG
CA80058-228EA 261.35 CAD
CA80058-228 CA80058-230 CA80058-232 CA80058-234 CA80058-236
HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid free acid ≥99% (by titrimetric analysis on dry substance), ULTROL®, Calbiochem®, Millipore®
HEPES (4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid)

A zwitterionic N-substituted aminosulfonic acid buffer. HEPES has its maximum buffering capacity very near the pH of mammalian tissue fluids (pKa of 7.31 at 37°C). Exhibits no metal binding and has a small temperature dependent ΔpKa of -0.014/°C.


  • Maximum buffering capacity
  • Useful in the pH 6.8 to 8.2 range
  • pKa (25 °C) 7.5


Absorbance (1.0 M, H₂O, 260 nm):≤0.05
Absorbance (1.0 M, H₂O, 280 nm): ≤0.04


Caution: Storage condition: OK to freeze

Formula: C₈H₁₈N₂O₄S
Storage Temperature: Ambient
MDL Number: MFCD00006158
CAS Number: 7365-45-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay ≥99% dry basis (titration)
Form Crystalline powder
Chloride (Cl-) ≤0.01%
Sulfate (SO42-) ≤0.05%
Color White


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR