PYREX® Culture Tubes
Supplier: Corning
9820-10 9820-13 9820-16 9820-16XX 9820-18 9820-20 9820-25 9820-25X
470211-658CS 969.65 CAD
470211-658 470211-662 470211-664 470211-668 470211-670 470211-672 470211-674 470211-676
PYREX® Culture Tubes
Tubes Culture Tubes
Made of PYREX® borosilicate glass, rimless.
  • Very good chemical resistance
  • Very good temperature resistance
  • Ends are fire-polished
Culture tubes have walls and bottom of a uniform thickness. All tubes feature a white marking spot, with the exception of 6 and 22 mm tubes.
Tube 5 ml can be used for Kahn tests. Tube 9 ml can be used for Wasserman tests.