Cleanert® SAX SPE Columns, Bonna-Agela
Supplier: Agela Technologies
SA0006 SA1001 SA2003 SA5003 SA5006
97018-226PK 1067.16 CAD
97018-226 97018-230 97018-234 97018-236 97018-238
Cleanert® SAX SPE Columns, Bonna-Agela
Chromatography Columns
Cleanert® SAX SPE products are packed with silica based sorbent bonded with a quaternary amine.
  • Suit to extraction of weak acids
  • Desalting of biological macromolecules
This strong anion exchanger is used to extract compounds capable of carrying a negative charge from both aqueous and non-aqueous solutions.