Uncharged Microscope Slides, Springside Scientific
Supplier: SPRINGSIDE SCIENTIFIC, LLC.
AB-90-WH AB-90-BL AB-90-PK AB-45-WH AB-45-BL AB-45-PK AB-90-WH-1B AB-90-BL-1B AB-90-PK-1B AB-45-WH-1B AB-45-BL-1B AB-45-PK-1B
75836-004CS 341.31 CAD
75836-004 75836-006 75836-008 75836-010 75836-012 75836-014 75836-154 75836-156 75836-158 75836-160 75836-162 75836-164
Uncharged Microscope Slides, Springside Scientific
Slides Microscope Slides
Microscope slides are manufactured from 3/4 white glass
These uncharged slides are standard size, and feature frosting optimized for hot foil tape labelers such as the Slidemate and Primera.
Ordering information: Slides are packed 72 per cardboard box. Each box is individually sealed. Each case contains 20 boxes.