Adhesive/Charged Microscope Slides, Springside Scientific
Supplier: SPRINGSIDE SCIENTIFIC, LLC.
AHS45-PK AHS90-WH-1B AHS90-BL01-1B AHS90-PK01-1B AHS45-WH01-1B AHS45-BL-1B AHS45-PK-1B AHS90-WH AHS90-BL AHS90-PK AHS45-WH AHS45-BL
75836-002CS 484.48 CAD
75836-002 75836-142 75836-144 75836-146 75836-148 75836-150 75836-152 75836-218 75836-220 75836-222 75836-224 75836-226
Adhesive/Charged Microscope Slides, Springside Scientific
Slides Adhesive Slides
Microscope slides are manufactured from 3/4 white glass and feature a positive charge for improved adhesion
All slides are standard size, with frosting optimized for hot foil tape labelers such as the Slidemate and Primera.
Ordering information: Slides are packed 72 per white cardboard box. Each box is individually sealed. Each case contains 20 boxes.