Tryptic Soy Broth (TSB)
Supplier: HARDY DIAGNOSTICS CA
K88
K285
CA89408-106PK
220.32
CAD
CA89408-106
CA10015-459
Tryptic Soy Broth (TSB)
Media for Microbiology
Dehydrated Media for Microbiology
Tryptic Soy Broth (TSB), filtered to remove dead bacteria.
Tryptic Soy Broth (TSB) is recommended for use as a general purpose medium for the isolation and cultivation of a wide variety of bacteria and fungi.
Ordering information: Minimum order of 5 packs.