Middlebrook 7H10, Deep Fill Plates
Supplier: HARDY DIAGNOSTICS CA
W30 C34
CA10015-460PK 116.81 CAD
CA10015-460 CA76006-346
Middlebrook 7H10, Deep Fill Plates
Media for Microbiology Dehydrated Media for Microbiology
Middlebrook 7H10 Agar is recommended for use in qualitative procedures for the isolation and cultivation of Mycobacterium species.
  • Optically clear polystyrene: For distortion free viewing
  • Heavy-duty: Reduces breakage during shipping
  • Stacking rings: Polystyrene petri plates feature raised rings for stacking ability and prevents sliding
  • Breathable packaging: Packaged in sleeves of ten plates in a breathable bag that prevents buildup of condensation and excess moisture
Industry: Clinical