Fluid A, USP
Supplier: HARDY DIAGNOSTICS CA
U436 U208
CA89407-529PK 689.46 CAD
CA89407-529 CA10124-490 CA89407-526
Fluid A, USP
Microbiology Test Systems Endotoxin Tests
A rinsing and diluting fluid for use in USP testing protocols.
A rinsing and diluting fluid for use in USP testing protocols. Contains peptone 0.1%.
Hardy Diagnostics' sterility media and rinse solutions are manufactured in an ISO 13485 environmentally controlled production center. Each lot is certified for pH, sterility and growth promotion using ATCC® strains specified by the USP. Our approach to manufacturing ensures all sterility media and rinse fluids provide the highest level of clarity for improved reading accuracy, thus significantly reducing the risk of incorrect interpretation and false results.
Ordering information: Order by the pack of 10