a1-3,4,6 Galactosidase, New England Biolabs
Supplier: New England Biolabs (NEB)
P0747L P0747S P0747L P0747S
102902-448EA 1000.14 CAD
102902-448 102902-450 CA102902-448 CA102902-450
a1-3,4,6 Galactosidase, New England Biolabs
Protein Purification Protein Purification Reagents
a1-3,4,6 Galactosidase is a highly specific exoglycosidase that catalyzes the hydrolysis of α1-3, α1-4 and α1-6 linked d-galactopyranosyl residues from oligosaccharides.