Nb.BssSI
Supplier: New England Biolabs (NEB)
R0681S R0681T R0681S R0681T
103218-948EA 129.56 CAD
103218-948 103218-950 CA103218-948 CA103218-950
Nb.BssSI
Enzymes
A restriction endonuclease that recognizes the sequence CACGAG.
  • Is a nicking endonuclease that cleaves only one strand of DNA on a dsDNA substrate
  • This is an Enzyme for Innovation (EFI)
Nicking Endonucleases can generate nicked or gapped duplex DNA for studies of DNA mismatch repair and for diagnostic applications. The long overhangs that nicking enzymes make can be used in DNA fragment assembly.