NEBuilder HiFi DNA Assembly Bundle for Large Fragments, New England Biolabs
Supplier: New England Biolabs (NEB)
NEBuilder HiFi DNA Assembly was developed to improve the efficiency and accuracy of DNA assembly
- Enjoy less screening/re-sequencing of constructs, with virtually error-free, high-fidelity assembly
- Join DNA fragments together more efficiently, even with larger fragments or low DNA inputs
- Use NEBuilder HiFi in successive rounds of assembly, because it removes 5' and 3' end mismatches
- Bridge two ds-fragments with a synthetic ss-DNA oligo for simple and fast construction
- Use with lower DNA input requirements
NEB recommends NEB 10-beta Competent E. coli, High Efficiency for assemblies larger than 15 kb. NEBuilder HiFi DNA Assembly Bundle for Large Fragments offers a discount for purchasing these competent cells with the NEBuilder HiFi DNA Assembly Master Mix. NEBuilder HiFi DNA Assembly Master Mix was developed to improve the efficiency and accuracy of DNA assembly. This method allows for seamless assembly of multiple DNA fragments, regardless of fragment length or end compatibility. This method has been used to assemble either single-stranded oligonucleotides or different sizes of DNA fragments with varied overlaps (15 to 30 bp). It has utility for the synthetic biology community, as well as those interested in one-step cloning of multiple fragments due to its ease of use, flexibility and simple master-mix format.