T3 RNA Polymerase, New England Biolabs
Supplier: New England Biolabs (NEB)
M0378S
M0378S
102902-444EA
132.72
CAD
102902-444
CA102902-444
T3 RNA Polymerase, New England Biolabs
Nucleic Acid Reagents
RNA Synthesis and Transcription
Bacteriophage T3 RNA Polymerase is a DNA-dependent RNA polymerase that is highly specific for the T3 phage promoters
The 99 KD enzyme catalyzes in vitro RNA synthesis from a cloned DNA sequence under the T3 promoters. RNA produced using the T3 RNA Polymerase is suitable for many applications in research and biotechnology.