NEBNext Ultra II Ligation Module, New England Biolabs
Supplier: New England Biolabs (NEB)
E7595L E7595S E7595L E7595S
102969-104EA 2150.38 CAD
102969-104 102969-106 CA102969-104 CA102969-106
NEBNext Ultra II Ligation Module, New England Biolabs
Nucleic Acid Reagents Cloning Kits and Ligation
The NEBNext Ultra II Ligation Module has been optimized to efficiently ligate DNA adaptors compatible with Illumina® sequencing to end-repaired, dA-tailed DNA fragments
The module is optimized for use with the NEBNext Ultra II End Repair/dA-Tailing Module (NEB #E7546), and is part of the Ultra II workflow which enables high yield preparation of high quality libraries from 500 picograms to 1 microgram of input DNA. It is also designed for use with NEBNext Ultra II Q5® Master Mix (NEB #M0544) and NEBNext Singleplex or Multiplex Oligos for Illumina (NEB #E7350, #E7335, #E7500, #E7600 or #E7535).