Isothermal Amplification Buffer II Pack, New England Biolabs
Supplier: New England Biolabs (NEB)
B0374S B0374S
102969-092EA 52.14 CAD
102969-092 CA102969-092
Isothermal Amplification Buffer II Pack, New England Biolabs
Nucleic Acid Reagents Isothermal Amplification Reagents
10X Isothermal Amplification Buffer II is optimized for use with Bst 3.0 DNA Polymerase
This buffer provides superior reaction conditions for isothermal amplification applications.