DNA (Deoxyribonucleic acid) (from salmon sperm)

Supplier: Rockland Immunochemical

MB-103-1000 MB-103-0025
CAMB-103-1000EA 294.01 CAD
CAMB-103-1000 CARLMB103-0025
DNA (Deoxyribonucleic acid) (from salmon sperm)
DNA (Deoxyribonucleic acid)

This product is a convenient, ready-to-use solution of salmon sperm DNA especially prepared for use in the preparation of pre-hybridization and hybridization solutions and as a DNA carrier in yeast transformation protocols and other related methods


This solution contains sheared single-stranded DNA molecules that can be used to block the non-specific attachment of probe DNA to the surface of a membrane (Southern) or to increase yeast transformation efficiency.

MDL Number: MFCD00130922
CAS Number: 9007-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR