SP Wilmad-LabGlass Volumetric Flasks with Plastic Stopper, Class A, SP Industries
Supplier: SP Industries
LG-8116-126 LG-8116-132 LG-8116-128
14201-494EA 74.09 CAD
14201-494 14202-050 14202-782 14203-310 14204-590 14205-558 14206-086 14206-136 14206-992 14207-000 14207-484 14208-774 14209-442 14210-620 14210-678 14211-012
SP Wilmad-LabGlass Volumetric Flasks with Plastic Stopper, Class A, SP Industries
Flasks Volumetric Flasks
These borosilicate glass volumetric flasks are provided with a plastic closed-bottom stopper which fits the Standard Taper ground zone
Stopper design prevents the neck from breaking if flask is tipped over. Calibrated "to contain" (TC) at 20°C.