Laundry Bag, Associated Bag
Supplier: Associated Bag
19-1-05 19-1-07
89087-548CS 386.67 CAD
89087-548 89087-552
Laundry Bag, Associated Bag
Bags Laundry Bags
Perfect for hospitality and healthcare industries, these laundry bags are water soluble.
  • Puncture and static - resistant
  • Attached tie tap for closure
  • Cost effective, puncture-resistant bags hold wet or damp linens
They won’t damage fabrics, equipment, or waste-water treatment systems. The bags are highly resistant to most organic solvents, greases, fats, and oils and are impervious to most gases. They won't generate a static charge.
Bags dissolve completely in water at 140 °F and fit 25" hamper.
Packaging: 100 per Carton.