Ultratorch® Solder Irons, Master Appliance
Supplier: Master Appliance
UT-100SI UT-40SI
94025-818EA 423.6 CAD
94025-818 94025-824
Ultratorch® Solder Irons, Master Appliance
Heaters Hot Air Dryers
The versatility in size and power of these Microtorch® butane torches offer something for everyone, and no matter what job you have on your “to do” list, we have the portable soldering iron and butane torches perfect for any task at hand.
  • Professional quality
  • Self-igniting
  • Butane powered soldering/heat tool and torch
  • Oxygen free, copper based Ultra tip soldering tips
  • Thick iron and chrome plating for long life and high thermal conductivity
  • Completely portable
  • Ready to use in 30-seconds
  • Adjustable temperature control
  • Patented gas catalyst reaction system
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Length 260 mm (10.3") 208 mm (8.3")