Corrugated SwitchBoard™ Anti-Fatigue Matting, Wearwell®
Supplier: Wearwell
702.38X3X75BK 702.14X2X75BK 702.14X3X75BK 702.14X4X75BK
89020-854RL 7929.91 CAD
89020-854 89021-394 89021-396 89021-398
Corrugated SwitchBoard™ Anti-Fatigue Matting, Wearwell®
Floor Mats Traction and Anti-Fatigue Floor Mats
Corrugated SwitchBoard™ matting prevents life-threatening electrical shocks in high-voltage environments.
  • Conforms to ANSI/ASTM D-178-01 Type II, Class 2
  • Dielectric strength: 30,000 volts
  • Recommended maximum usage per ASTM D178-01: up to 17,000 volts
  • Easy to clean corrugated surface design
Designed to protect personnel in areas with dangerous high-voltage equipment. Surface is easily cleaned and will not collect contaminants. Conforms to ANSI/ASTM D-178-01 Type II (a premium quality vinyl compound resistant to UV, ozone and many common chemicals).More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable