Hydrochloric acid 10% w/v, EMPROVE® EXPERT NF, JP, Ph. Eur., SAFC®

Supplier: EMD MILLIPORE – PCS CA EXCEP
Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

4.80592.9026 4.80592.9200 4.80592.9950
CA4.80592.9026EA 1078.92 CAD
CA4.80592.9026 CA4.80592.9200 CA4.80592.9950
Hydrochloric acid 10% w/v, EMPROVE® EXPERT NF, JP, Ph. Eur., SAFC®
Hydrochloric acid

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated.


  • Filtered with 0.2 µm


Certifications: Corresponds to Ph. Eur., JP, NF.

Formula: HCl
MW: 36.46
Boiling Pt: 102 °C
Density: 1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

Assay (alkalimetric) 10.0 - 10.5 % (w/v)
Identity passes test
Color ≤ 10 Hazen
Appearance passes test
Bromide or iodide passes test
Free bromine or chlorine passes test
Free chlorine (Cl) ≤ 0.0001
Sulfate (SO₄) ≤ 0.0005 %
Sulphite (SO3) ≤ 0.001 %
Heavy metals (as Pb) ≤ 0.0001 %
As (Arsenic) ≤ 0.000015 %
Hg (Mercury) ≤ 0.0000010 %
Residual solvents (ICH Q3C) ...excluded by production process
Evaporation residue ≤ 0.01
Residue on ignition (as SO₄) ≤ 0.008 %
Endotoxins ≤ 2.5 I.U./ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR