Adenosine-5'-triphosphate disodium salt (ATP disodium salt) trihydrate ≥98% (by HPLC)
Supplier: CHEM-IMPEX INTERNATIONAL, INC
Synonyms:
(((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl)triphosphoric acid, disodium salt trihydrate, 5'-ATP·2Na·3H2O, Adenosine-5'-triphosphoric acid disodium salt trihydrate, ATP, 2Na 3H2O, ATP disodium trihydrate
00015-100G
00015-25G
CH00015-100GEA
142.2
CAD
CH00015-100G
CH00015-25G
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) trihydrate ≥98% (by HPLC)
Adenosine 5'-triphosphate disodium salt (ATP disodium salt) trihydrate
Formula:
C₁₀H₁₄N₅Na₂O₁₃P₃·3H₂O MW: 605.19 g/mol Melting Pt: 176 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00150754 CAS Number: 51963-61-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...