Difco™ Cooke Rose Bengal Agar, BD
Supplier: BD Life Sciences
270310 270310
CA90003-836EA 405.87 CAD
CA90003-836 90003-836
Difco™ Cooke Rose Bengal Agar, BD
Media for Microbiology Dehydrated Media for Microbiology
Cooke Rose Bengal Agar is used for selective cultivation and isolation of fungi.
  • Papaic digest of soybean meal provides nitrogen, carbon, and vitamins
  • Dextrose is an energy source
  • Rose bengal and chlortetracycline inhibit bacterial growth
  • Monopotassium phosphate acts as a buffering agent
  • Magnesium sulfate is a source of divalent cations
The medium should not be exposed to light, as photo-degradation of rose bengal yields compounds that are toxic to fungi.