Microtorch® Butane Powered Torches, Master Appliance
Supplier: Master Appliance
MT-51 MT-76 GT-70 MT-11
94025-820CS 1166.06 CAD
94025-820 94025-822 75845-486 75845-488
Microtorch® Butane Powered Torches, Master Appliance
Heaters Hot Air Dryers
Master butane torches are perfect for any task at hand. They are self-contained, butane-powered, heavy-duty, general industrial torches.
  • Self-igniting piezo electric ignition that is easy to start and eliminates need for lighter
  • Self-contained, metal fuel tank that is rugged and lightweight, easy to use, eliminates bulky canister
  • Large fuel capacity, 80 cc self-contained tank for 30-minute continuous run time
  • Large sized adjustable blue flame that accommodates many heavy-duty industrial applications with temperatures up to 2500 °F/1300 °C
  • Safety lock that
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable