Back-Loading Printed Press-on Envelope, Associated Bag
Supplier: Associated Bag
45-3-9522 45-2-14
80085-666CS 673.41 CAD
80085-666 89140-080
Back-Loading Printed Press-on Envelope, Associated Bag
Envelopes
This envelope sticks to packages to protect shipping documents from dirt and moisture.
Back-loading style forms tamper-evident pocket for papers
Pressure-Sensitive Adhesive
Dimension-Listed Open End
"Packing List Enclosed" Printed on Envelope
Tamper-Evident Pocket
The pocket must be torn or cut open to retrieve papers.
Pressure-sensitive adhesive sticks aggressively to paper, metal, wood, corrugated cardboard.