Anti-F4/80 Rat Monoclonal Antibody (redFluor™ 710) [clone: BM8.1]

Supplier: Tonbo Biosciences
80-4801-U025 80-4801-U100
75834-374EA 219.62 CAD
75834-374 75834-376
Anti-F4/80 Rat Monoclonal Antibody (redFluor™ 710) [clone: BM8.1]
Antibodies
The BM8.1 antibody is specific for mouse F4/80 antigen, a 125 kDa transmembrane protein widely expressed by members of the mononuclear phagocyte system and considered to be a key marker for mature macrophage cells. F4/80 is differentially expressed during myeloid cell development, and may be regulated by certain cytokines within the tissue microenvironment. Other cell types shown to express this antigen include Langerhans cells, Kupffer cells and dendritic cell subsets. BM8.1 is widely used together with antibodies to CD115 (c-fms), CD11b and CD11c to identify myeloid / macrophage cells by flow cytometry.

Type: Primary
Antigen: F4/80
Clonality: Monoclonal
Clone: BM8.1
Conjugation: redFluor™ 710
Epitope:
Host: Rat
Isotype: Rat IgG2b, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR