Anti-CD20 Mouse Monoclonal Antibody (APC-Cyanine7) [clone: 2H7]

Supplier: Tonbo Biosciences
25-0209-T025 25-0209-T100
75834-362EA 219.62 CAD
75834-362 75834-364
Anti-CD20 Mouse Monoclonal Antibody (APC-Cyanine7) [clone: 2H7]
Antibodies
The 2H7 antibody reacts with human CD20, expressed by pre-B cells, resting and activated B cells, follicular dendritic cells and a small subset of T cells, but not on plasma cells. CD20 is a 33-37 kDa transmembrane protein which is differentially phosphorylated in resting and active B cells. The CD20 molecule plays a role in regulating B-cell activation. The 2H7 antibody is reported to be cross-reactive with several non-human species including Baboon, Chimpanzee, Cynomolgus and Rhesus. Please choose the appropriate format for each application.

Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: 2H7
Conjugation: APC-Cyanine7
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR