SP Wilmad-LabGlass Cylindrical Reaction Vessels with Flat Flange, SP Industries
Supplier: SP Industries
LG-8080-102 LG-8080-100
14207-638EA 670.11 CAD
14207-638 14207-656
SP Wilmad-LabGlass Cylindrical Reaction Vessels with Flat Flange, SP Industries
Reaction Vessels Reaction Vessels, Cylindrical Shape
These borosilicate glass cylindrical reaction vessels are without constriction at the top and have a flat bottom.
  • Sturdy walls
  • Tooled flat flange
They feature sturdy walls and a tooled flat flange with a surface ground to assure a tight seal. There is a gently sloping contour at junction of side wall and bottom, thus preventing any weakening that might occur with a sharp shoulder.
Ordering information: Uses LG-7316-106 clamp.