Climate Chambers ICH Series, Memmert
Supplier: Memmert
ICH110115V ICH110230V ICH260115V ICH260230V ICH750115V ICH750230V
10861-360EA 23996.43 CAD
10861-360 10861-362 10861-364 10861-366 10861-368 10861-370
Climate Chambers ICH Series, Memmert
Testing Chambers Climatic Chambers
Memmert stability chambers are constructed of stainless steel inside and out. The ICH L stability chamber will save energy and perform well for any project.
  • From temperatures of at least 10 up to +60 °C including air jacket heating system for gentle all-around heating
  • Humidity range 10 to 80% rh - or rh OFF
  • Illumination unit consisting of 3 fluorescent lights with cold white light (standard illuminant D65, 6.500K) and 2 UV-lamps in the spectral range of 320-400 nm
  • Programmable with internal data logging via included AtmoCONTROL Software
  • Available in 3
More Product Information
Order Now
SPECIFICATIONS
Temperature range –10 °C up to 60 °C without humiditiy; 10 °C up to 60 °C with humidity
Environmentally Preferable Sustainable Packaging,Low Manufacturing Impact
Control range Temperature range: Min 5 °C above ambient up to 50 °C; Humidity range: 40 to 97 % rh (and rh Off option)
Environmental certification EU Directives 2002/95/EC (RoHS) and 2002/96/EC (WEEE)