Peltier Cooled Incubators, IPP, Memmert
Supplier: Memmert
IPP55PLUS115V IPP30PLUS115V IPP30115V IPP55115V
10861-432EA 10039.04 CAD
10861-432 10861-460 10861-468 10861-328 10861-330
Peltier Cooled Incubators, IPP, Memmert
Incubators Cooled Incubators
Peltier Cooled Incubator IPP* with Single DISPLAY
PELTIER COOLED INCUBATOR IPP Heating and cooling seamlessly with one system thanks to Peltier technology. In this respect, cooled incubators IPP not only contribute to climate protection, but it also achieves an additional decrease in operating costs of up 90% compared to compressor technology. This perfect development from the environmentally friendly and energy-saving heating/cooling technology by Memmert convinces by outstanding control precision and extremely small fluctuations.
Peltier Cooled Incubator IPPplus* with Twin DISPLAY. AtmoCONTROL Software.
Certifications: CE Marked.