Determination of Water Hardness

Supplier: ALDON CORP SE
IS9751
470146-438EA 80.05 CAD
470146-438
Determination of Water Hardness
Educational Materials Chemistry Educational Materials Mixtures and Solutions Learning Activities
Use Color Transition of Titration to Determine Water Hardness.

  • Great for field studies
  • Perfect environmental or chemistry courses
  • All chemicals required are included

This kit has been designed to allow the analyst to perform a very simple titration to determine the hardness of their fresh water sample. The titration uses an easily identifiable color transition to indicate the end of the analysis. The kit contains everything an analyst would need to perform a water analysis either in the laboratory setting or out in the field. Kit contains enough materials to perform 40 tests.

Includes:
Plastic Pipettes
Reaction Vessel
Collection Bottle
Measuring Tube
Titrator
EDTA Titrant
pH 10 Buffer Solution
Ericochrome Black T Indicator Solution
Teacher's Guide and Student Copymasters
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR