Telescoping Base Stands for Purifier® Horizontal Clean Bench, Labconco®
Supplier: Labconco
3888700 3888701 3613200 3613201 3857700 3857701 3857300 3857301 3255000 3255001
89203-906EA 2395.44 CAD
89203-906 89203-908 89232-326 89232-328 89402-690 89402-692 89402-694 89402-696 89497-262 89497-264
Telescoping Base Stands for Purifier® Horizontal Clean Bench, Labconco®
Retort Stands
These stands have telescoping legs so that they may be adjusted to eight height positions in 1" increments from 27.5" to 34.5"
  • Stands are available with fixed leveling feet or casters for mobility
Frame is constructed of durable, 1.75" tubular, glacier white powder-coated steel. Rear crossbar supports offer additional stability. Support loads up to 1000 pounds. ADA-compliant. Casters are 5" diameter, toe-locking, non-marking polyurethane casters with bearings for quiet operation.
Ordering information: Designed for use with Labconco Purifier Horizontal Clean Benches. Height selection and adjustment must be completed before the enclosure is installed atop the stand.