Superwhite Coverglass, Springside Scientific
Supplier: SPRINGSIDE SCIENTIFIC, LLC.
HCW122450 HCW122455 HCW122460
75836-078CS 51.11 CAD
75836-078 75836-080 75836-082
Superwhite Coverglass, Springside Scientific
Cover Glasses
Microscope coverslips are made of pure, super white glass which prevents auto-fluorescence
They work well in all automated coverslippers with no sticking or breaking. Varied sizes are available.
Ordering information: Each plastic box contains 1oz. Each case includes 10 boxes, and is vaccum sealed.