Inkjet Superwhite Microscope Slides, Springside Scientific
Supplier: SPRINGSIDE SCIENTIFIC, LLC.
HMW202F9W-1B HMW202F9Y-1B HMW202F9P-1B HMW232F9W HMW232F9Y HMW232F9P HMW202F9W HMW202F9Y HMW202F9P HMW232F9W-1B HMW232F9Y-1B HMW232F9P-1B
75836-046CS 30.34 CAD
75836-046 75836-048 75836-050 75836-066 75836-068 75836-070 75836-072 75836-074 75836-076 75836-202 75836-204 75836-206
Inkjet Superwhite Microscope Slides, Springside Scientific
Slides Microscope Slides
Inkjet Superwhite Microscope Slides are made of pure, super white glass which prevents auto-fluorescence
They are uniformly 75x25x1mm. Colored frosting is optimized for Leica inkjet labelers, ensuring consistently good barcode reads.
Ordering information: Charged slides are packaged in plastic boxes. Uncharged slides are packaged in ardboard boxes. Each box contains 72 slides, along with fiber desiccant. Each box is individually sealed. Each case contains 20 boxes.