Airfree® Adapters, Solid Addition, Chemglass
Supplier: Chemglass
AF-0503-01 AF-0503-02
80064-486EA 163.32 CAD
80064-486 80064-488
Airfree® Adapters, Solid Addition, Chemglass
Adapters Adapters, Conical Ground Joints
This adapter permits the addition of solids into multi-neck flasks in a dry box, or the addition of non-air sensitive compounds into flasks while purging with an inert atmosphere.
  • Use for solid addition
  • With drip tube
The adapter can handle from 50 mg to 1 g of solids. It is made with a drip tube to avoid getting the solids on the joint or the wall of the flask.