Microflex® 93-260 Gloves, Ansell
Supplier: Microflex
93260060 93260070 93260080 93260090 93260100 93260110
CA75832-934BX 34.97 CAD
CA75832-934 CA75832-936 CA75832-938 CA75832-940 CA75832-942 CA75832-944
Microflex® 93-260 Gloves, Ansell
Gloves
Disposable chemical resistant gloves.
  • Three layer design for superior protection against harsh chemicals including acids, solvents, and bases
  • Thin mil construction provides enhanced tactility and dexterity
  • Extra-soft material and ergonomic design provide outstanding fit, feel, and flexibility for longer wear time
  • Lower acceptable pinhole rate (0.65 AQL) and extended cuff for reliable protection against hazardous substances
  • Silicone-free formulation and processing ensure better product protection
  • For single use only
The Microflex® 93-260 series glove'sMore Product Information