Histology Control Slides, Trichrome, Springside Scientific
Supplier: SPRINGSIDE SCIENTIFIC, LLC.
CS-TCH20 CS-TCH100
75836-112CS 362.01 CAD
75836-112 75836-138
Histology Control Slides, Trichrome, Springside Scientific
Slides Microscopy Control Slides
100% Human Tissue.
Two stained controls in every box. Top-quality, pure white glass slides/coverglass.
Ordering information: Packaged 22 (20 controls plus 2 stained) or 100 (98 controls plus 2 stained) slides in a 25-postion or 100-position plastic slide-box.