Histology Control Slides, Histopath Fungus, Springside Scientific
Supplier: SPRINGSIDE SCIENTIFIC, LLC.
CS-HPF20 CS-HPF100
75836-096CS 402.43 CAD
75836-096 75836-122
Histology Control Slides, Histopath Fungus, Springside Scientific
Slides Microscopy Control Slides
100% Human Tissue.
Two stained controls in every box. Top-quality, pure white glass slides/coverglass.
Ordering information: Packaged 22 (20 controls plus 2 stained) or 100 (98 controls plus 2 stained) slides in a 25-postion or 100-position plastic slide-box.