Histology Control Slides, Helico Bacteria, Springside Scientific
Supplier: SPRINGSIDE SCIENTIFIC, LLC.
CS-HLB20 CF-HLB100
75836-094CS 396.79 CAD
75836-094 75836-120
Histology Control Slides, Helico Bacteria, Springside Scientific
Slides Microscopy Control Slides
100% Human Tissue.
Two stained controls in every box. Top-quality, pure white glass slides/coverglass.
Ordering information: Packaged 22 (20 controls plus 2 stained) or 100 (98 controls plus 2 stained) slides in a 25-postion or 100-position plastic slide-box.