80 Octane Blend PRF, B&J Brand™ ASTM D2699/D2700, Burdick & Jackson™
Supplier: HONEYWELL BURDICK & JACKSON CA
|
Danger
|
RS2175-19
RS2175-204
BJRS2175-19EA
4431.46
CAD
BJRS2175-19
BJRS2175-204
80 Octane Blend PRF, B&J Brand™ ASTM D2699/D2700, Burdick & Jackson™
80 Octane Blend
Primary Reference Fuels
- Octane number, the principal performance characteristic of gasoline, is determined through use of certified PRF fuels and a Cooperative Fuel Research (CFR) engine under controlled test conditions
- used in both research and analytical applications
- thorough quality control testing to ensure each lot conforms to the specifications defined in ASTM D2699 for Motor Octane Number (MON) and ASTM 2700 for Research Octane Number (RON).
Certifications: Honeywell certifies that this product meets the requirements as defined in ASTM D2699 and D2700.
Boiling Pt:
6.7…7.0 %(V) Melting Pt: –107…–90 °C Density: 0.6901 g/cm³ (20 °C) Flash Pt: –12.2 °C Storage Temperature: Ambient |
CAS Number:
68920-06-9 UN: 1993 ADR: 3,II |
Specification Test Results
Appearance | Clear, colorless liquid |
Color (APHA) |
|
Water (Karl Fischer) |
|
Residue |
|
Composition by GC: Heptane | 19.9 – 20.1% |
Composition by GC: Iso-Octane | 79.9 – 80.1% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...