Sodium chloride solution_SC 0,9% USP, sterile (Normal saline)
Supplier: Cytiva
Synonyms:
Saline, NaCl, Sea salt, Halite
Z1377
Z1376
Z1377
Z1382
75832-226CS
271.92
CAD
75832-226
75832-228
CA75832-228
CA76081-514
CA75832-226
CA76881-882
Sodium chloride solution_SC 0,9% USP, sterile (Normal saline)
Sodium chloride
Sodium chloride injection is a sterile solution of sodium chloride in water for injection. It contains no antimicrobial agents. It contains NLT 95.0% and NMT 105.0% of the labeled amount of sodium chloride (NaCl). Manufactured in accordance with cGMP quality systems.
- Sterile and endotoxin tested making the product ideal for most research, testing and manufacturing applications including cell culture and molecular protocols
- Multiple packaging sizes available, including 200 l drum, 100 l drum, 20 l bag and 1 l bottle
Formula:
NaCl MW: 58.44 g/mol Boiling Pt: 1413 Melting Pt: 801 |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 Merck Index: 14,08599 |
Specification Test Results
Appearance | Clear Liquid (Visual) |
Chloride Identification | Pass |
Concentration | 0.90% |
Conductivity | 15.45 mS/cm |
Endotoxin |
4.5 - 7.0 (USP |
Iron | Pass |
NaCl | 8.99 g/L |
Particulate Matter | Meets USP Requirements (USP <788>) |
pH | 4.5 - 7.0 (USP <791>) |
Sodium Identification | Pass |
Sterility | No Growth (USP <71>) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...