Protease Inhibitor Cocktail Set I, Animal-Free, Calbiochem®, MilliporeSigma
Supplier: MilliporeSigma
535142-1ML 535142-1SET
CA80017-340EA 153.78 CAD
CA80017-340 CA80017-342
Protease Inhibitor Cocktail Set I, Animal-Free, Calbiochem®, MilliporeSigma
Cell Culture Supplements and Additives
Cocktail of five protease inhibitors for the inhibition of a broad range of proteases and esterases
Lyophilized. Reconstitute each vial with 1 mL dH2O to yield a 100X stock solution.