Tamperproof Specimen Containers, Simport Scientific
Supplier: Simport Scientific
C581-2600 C581-5700
CA89415-174CS 137.43 CAD
CA89415-174 CA89415-176
Tamperproof Specimen Containers, Simport Scientific
Sample Containers General Purpose Containers
Container and closure are made of polypropylene.
  • Tight-fitting lids prevent leaks and odors
  • Resist temperatures up to 121 °C
These disposable containers are ideal for collection, transport and storage of fluids, powders, solids, pathology specimens and hazardous samples. They incorporate a tamperproof lid which is opened by first removing tab on side of lid, then by lifting the cover.
Order Now
SPECIFICATIONS
Environmentally Preferable