XPR Microbalances, METTLER TOLEDO®
Supplier: Mettler Toledo
30279285 30279296 30279267 30279196 30279276
75832-778EA 60500 CAD
75832-778 75832-780 75832-782 75832-842 75832-844
XPR Microbalances, METTLER TOLEDO®
Balances and Scales
METTLER TOLEDO’s new XPR microbalances incorporate a range of innovative new technologies to deliver the highest accuracy in their class and help make your micro-weighing tasks simpler, safer and more secure.
  • Capacities from 2.1 g to 10.1 g
  • Readabilities from 0.1µg
  • Glass draft shield with motorized drive for precise weighing even in unstable environments.
  • Compact design with small footprint.
  • Fully automatic adjustment using internal weights.
  • Built-in level sensor and leveling aid for fast and easy leveling.
  • Capacitive color TFT touch screen
  • Two-terminal concept simplifies operation, with the the SmartView terminal sitting above the weighing chamber
  • LevelControl: Level warning with graphical levelling guide
  • Two programmable sensors for hands-free operation of the draft shield (SmartSens) to speed up frequently recurring tasks.
  • User management, change history function, and password protection.
  • Storage of SOPs in method library, including sample series and tolerances.
To make the most of your valuable resources, XPR microbalances and ultra-microbalances deliver a unique level of precision with exceptionally low minimum weights.
Certifications: UL listed and CSA certified.
Ordering information: Service packages are offered to assure continual reliability and maintain optimum balance performance. Proper installation and initial calibration are the key to ensuring the balance performs properly from day one. A METTLER TOLEDO technician performs installation qualification and operational qualification (IQ/OQ). Contact your representative for Service package details.